Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife Dry 1.5 mm, 35° Knife angle
SDP
Catalog No. 501902391
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 1.5 mm
Knife angle: 35°
Catalog No. 50-190-2391 Supplier Electron Microscopy Sciences Supplier No. 15ULD
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 1.5 mm
Knife angle: 35°