Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife Diamond Knife Dry, 2.4 mm, 35° Knife angle
SDP
Catalog No. 501902400
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 2.4 mm
Knife angle: 35°
Catalog No. 50-190-2400 Supplier Electron Microscopy Sciences Supplier No. 24ULD
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 2.4 mm
Knife angle: 35°