Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamedix™ 96-Well Assay Plates
SDP

Catalog No. 23451056
Encompass
Change view
Click to view available options
For Use With (Application):
Diamedix C6 Lyme Test Kit
Diamedix Cardiolipin, B2-Glycoprotein and Tissue Transglutaminase Test Kits
Diamedix Kits (other than Cardiolipin, B2-Glycoprotein, Tissue Transglutaminase, C6 Lyme)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
23-451-056 Diamedix C6 Lyme Test Kit
23-451-055 Diamedix Cardiolipin, B2-Glycoprotein and Tissue Transglutaminase Test Kits
23-451-054 Diamedix Kits (other than Cardiolipin, B2-Glycoprotein, Tissue Transglutaminase, C6 Lyme)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-451-056 Supplier Diamedix™ Supplier No. 250041
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

96-well pates to be used with Diamedix test kits

  • Blank well plates for use with Diamedix test kits
  • Each plate contains 96 wells
  • Fisher Cat No. 23-451-055 is meant to be used with Diamedix Cardiolipins, Diamedix β2-Glycoproteins and tissue Transglutaminase test kits
  • Fisher Cat No. 23-451-056 is meant to be used with Diamedix C6 Lyme test kit

Specifications

Format Standard
Array 8 x 12
Product Type Microplate Blank-Well
Material Polystyrene
No. of Wells 96
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
For Use With (Application) Diamedix C6 Lyme Test Kit
Quantity 2/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.