Learn More
Diamedix™ Immunosimplicity™ anti-dsDNA Test Kit
SDP
Catalog No. 23451009
Encompass
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
23-451-009 96 Tests
1 options
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Catalog No. 23-451-009 Supplier Diamedix™ Supplier No. 720700
Add to Cart
Add to Cart
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes
Antigen Wells, 0 IU/mL Standard, 12.5 IU/mL Standard, 25 IU/mL Standard, 50 IU/mL Standard, 100 IU/mL Standard, 200 IU/mL Standard, Positive Control, Negative Control, Sample E Diluent, Wash U Concentrate (20X), Conjugate, Substrate HRP, Stop O Solution
Detects IgG antibodies to double-stranded (ds)DNA in human serum
The Diamedix™ Immunosimplicity™ anti-dsDNA Test Kit is a quantitative indirect enzyme immunoassay
  • Detects antibodies against the dsDNA antigen in human serum
  • Aids in the diagnosis of systemic lupus erythematosus (SLE)
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Detectable Analytes dsDNA IgG
Format 96-well Plate
Type Anti-dsDNA Test Kit
Includes Antigen wells, 0 IU/mL standard, 12.5 IU/mL standard, 25 IU/mL standard, 50 IU/mL standard, 100 IU/mL standard, 200 IU/mL standard, positive control, negative control, sample E diluent, wash U concentrate 20X, conjugate, substrate HRP, stop O solution
Quantity 96 Tests