Learn More
Diamedix™ Immunosimplicity™ anti-MPO (P-ANCA) Test Kit
SDP
Catalog No. 23451017
Encompass
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
23-451-017 96 Tests
1 options
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Catalog No. 23-451-017 Supplier Diamedix™ Supplier No. 720430
Add to Cart
Add to Cart
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes
Antigen Wells, 0 EU/mL Standard, 5 EU/mL Standard, 12.5 EU/mL Standard, 25 EU/mL Standard, 50 EU/mL Standard, 100 EU/mL Standard, Low Positive Control, Negative Control, Sample A Diluent, Wash T Concentrate (20X), Conjugate, Substrate HRP, Stop N Solution
Detects antibodies against the myeloperoxidase (MPO) antigen in serum
The Diamedix™ Immunosimplicity™ anti-MPO (P-ANCA) Test Kit is a semi-quantitative and qualitative enzyme immunoassay
  • Detects antibodies against the myeloperoxidase (MPO) antigen in serum
  • Aids in the diagnosis of Wegener's granulomatosis
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Detectable Analytes Proteinase-3 (PR-3) Antibodies
Format 96-well Plate
Type Anti-MPO (P-ANCA) Test Kit
Includes Antigen wells, 0 EU/mL standard, 5 EU/mL standard, 12.5 EU/mL standard, 25 EU/mL standard, 50 EU/mL standard, 100 EU/mL standard, low positive control, negative control, sample A diluent, wash T concentrate 20X, conjugate, substrate HRP, stop N solution
Quantity 96 Tests