Learn More
Diamedix™ Immunosimplicity™ anti-Sm/RNP Test Kit
SDP
Catalog No. 23451006
Encompass
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
23-451-006 96 Tests
1 options
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Catalog No. 23-451-006 Supplier Diamedix™ Supplier No. 720270
Add to Cart
Add to Cart
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes
Antigen Wells, Calibrator, Postive Control, Negative Control, Sample Diluent, Wash Concentrate (20X), Conjugate, Substrate, Stop Solution
Detects antibodies against the Sm/RNP antigen in serum
The Diamedix™ Immunosimplicity™ anti-Sm/RNP Test Kit is a semi-quantitative enzyme immunoassay
  • Detects antibodies against the Sm/RNP antigen in serum
  • Aids in the diagnosis of autoimmune disease
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Detectable Analytes SSA (Ro) Antibodies
Format 96-well Plate
Type Anti-Sm/RNP Test Kit
Includes Antigen wells, calibrator, positive control, negative control, sample diluent, wash concentrate 20X, conjugate, substrate, stop solution
Quantity 96 Tests