Learn More
Diamedix™ Immunosimplicity™ anti-Sm Test Kit
SDP
Catalog No. 23451005
Encompass
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
23-451-005 96 Tests
1 options
This item is not returnable. View return policy
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Catalog No. 23-451-005 Supplier Diamedix™ Supplier No. 720240
Add to Cart
Add to Cart
This item is not returnable. View return policy
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes
Antigen Wells, Calibrator, Postive Control, Negative Control, Sample Diluent, Wash Concentrate (20X), Conjugate, Substrate, Stop Solution
Detects antibodies against the Sm antigen in serum
The Diamedix™ Immunosimplicity™ anti-Sm Test Kit is a semi-quantitative enzyme immunoassay
  • Detects antibodies to the Sm antigen in serum
  • Aids in the diagnosis of autoimmune disease
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Detectable Analytes Sm Antibodies
Format 96-well Plate
Type Anti-Sm Test Kit
Includes Antigen wells, calibrator, positive control, negative control, sample diluent, wash concentrate 20X, conjugate, substrate, stop solution
Quantity 96 Tests