Learn More
Diamedix™ Immunosimplicity™ anti-SSB Test Kit
SDP
Catalog No. 23451004
Encompass
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
23-451-004 96 Tests
1 options
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Catalog No. 23-451-004 Supplier Diamedix™ Supplier No. 720260
Add to Cart
Add to Cart
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes
Antigen Wells, Calibrator, Postive Control, Negative Control, Sample Diluent, Wash Concentrate (20X), Conjugate, Substrate, Stop Solution
Detects antibodies against the SSB (La) antigen in serum
The Diamedix™ Immunosimplicity™ anti-SSB Test Kit is a semi-quantitative enzyme immunoassay
  • Detects antibodies against the SSB (La) antigen in serum
  • Aids in the diagnosis of autoimmune disease
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Detectable Analytes SS-B/La Antibodies
Format 96-well Plate
Type Anti-SSB Test Kit
Includes Antigen wells, calibrator, positive control, negative control, sample diluent, wash concentrate 20X, conjugate, substrate, stop solution
Quantity 96 Tests