Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamedix™ Immunosimplicity™ anti-TPO Test Kit
SDP

Catalog No. 23451019
Encompass
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-451-019 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Handle samples, standards, controls and the materials that contact them as potential biohazards.

Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.

Catalog No. 23-451-019 Supplier Diamedix™ Supplier No. 720450
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Handle samples, standards, controls and the materials that contact them as potential biohazards.

Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.

Includes

Antigen Wells, Standards: 0 IU/mL, 25 IU/mL, 50 IU/mL, 100 IU/mL, 300 IU/mL, 1000 IU/mL, Low Positive Control, Negative Control, Sample A Diluent, Wash T Concentrate (20X), Conjugate, Substrate HRP, Stop N Solution

Detects antibodies against the thyroid peroxidase (TPO) antigen in serum

The Diamedix™ Immunosimplicity™ anti-TPO Test Kit is a quantitative and qualitative enzyme immunoassay

  • Detects and quantifies antibodies to thyroid peroxidase antigen in serum
  • Aids in the diagnosis of autoimmune thyroid disease
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems

Specifications

Detectable Analytes Thyroid Peroxidase Autoantibodies
Format 96-well Plate
Type Anti-TPO Test Kit
Includes Antigen wells, standards: 0 IU/mL, 25 IU/mL, 50 IU/mL, 100 IU/mL, 300 IU/mL, 1000 IU/mL, low positive control, negative control, sample A diluent, wash T concentrate 20X, conjugate, substrate HRP, stop N solution
Quantity 96 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.