Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamedix™ Immunosimplicity™ EBV-VCA IgG Test Kit
SDP

Catalog No. 23451029
Encompass
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-451-029 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Handle samples, calibrators, controls and the materials that contact them as potential biohazards.

Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.

Catalog No. 23-451-029 Supplier Diamedix™ Supplier No. 720600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Handle samples, calibrators, controls and the materials that contact them as potential biohazards.

Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.

Includes

Antigen Wells, Cut-off Calibrator, Low Positive Control, Negative Control, Sample B Diluent, Wash T Concentrate (20X), Conjugate, Substrate HRP, Stop N Solution

Detects IgG antibodies to Epstein-Barr Virus Viral Capsid Antigen in human serum

The Diamedix™ Immunosimplicity™ EBV-VCA IgG Test Kit is a qualitative and semi-quantitative indirect enzyme immunoassay

  • Detects IgG antibodies to Epstein-Barr Virus (recombinant) Viral Capsid Antigen (EBV-VCA) in human serum
  • May be used in combination with other Epstein-Barr serologies, Viral Capsid Antigen (VCA) IgM, Epstein-Barr Nuclear Antigen-1 (EBNA-1) IgG and IgM, Early Antigen-Diffuse (EA-D) IgG and IgM, and heterophile antibody as an aid in the diagnosis of infectious mononucleosis (IM)
  • The evaluation of paired sera, to determine a significant increase in VCA IgG antibody titer, can also aid in the diagnosis of acute infection
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems

Specifications

Detectable Analytes EBV-VCA IgG
Format 96-well Plate
Type Immunosimplicity™ EBV-VCA IgG Test Kit
For Use With (Application) Qualitative and semi-quant determination of IgG Antibodies to EBV (recombinant) VCA
Includes Antigen wells, cut-off calibrator, low positive control, negative control, sample B diluent, wash T concentrate 20X, conjugate, Substrate HRP, Stop N Solution
Quantity 96 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.