Learn More
Diamedix™ Immunosimplicity™ ENA-6 Screen
SDP
Catalog No. 23451002
Encompass
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
23-451-002 96 Tests
1 options
This item is not returnable. View return policy
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Catalog No. 23-451-002 Supplier Diamedix™ Supplier No. 720230
Add to Cart
Add to Cart
This item is not returnable. View return policy
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes
Antigen Wells, Cut-off Calibrator, Low Positive Control, Negative Control, Sample Diluent, Wash Concentrate (20x), Conjugate, Substrate, Stop Solution
For the detection of IgG antibodies to six ENAs in human serum
The Diamedix™ Immunosimplicity™ ENA-6 Screen Test Kit is a qualitative indirect enzyme immunoassay
  • Detects IgG antibodies to six extractable nuclear antigens (ENAs) in human serum
  • Aids in the diagnosis of certain autoimmune disorders
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
One well can be used to screen for antibodies to:
  • Sm
  • Sm/RNP
  • SSA
  • SSB
  • Scl-70
  • Jo-1
Specifications
Detectable Analytes Human IgG
Format 96-well Plate
Type ENA-6 Screen
Includes Antigen wells, cut-off calibrator, low positive control, negative control, sample C diluent, wash T concentrate 20X, conjugate, substrate HRP, stop N solution
Quantity 96 Tests