Learn More
Diamedix™ Immunosimplicity™ Mumps IgG Test Kit
SDP
Catalog No. 23451037
Encompass
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
23-451-037 96 Tests
1 options
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Catalog No. 23-451-037 Supplier Diamedix™ Supplier No. 720540
Add to Cart
Add to Cart
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes
Antigen Wells, Cut-off Calibrator, Low Positive Control, Negative Control, Sample B Diluent, Wash T Concentrate (20X), Conjugate, Substrate HRP, Stop N Solution
Detects IgG antibodies to mumps virus in human serum
The Diamedix™ Immunosimplicity™ Mumps IgG Test Kit is a qualitative and semi-quantitative indirect enzyme immunoassay
  • Detects IgG antibodies to mumps virus in human serum
  • Aids in the diagnosis of Mumps infection
  • The evaluation of paired sera, to determine a significant increase in Mumps IgG antibody titer, can also aid in the diagnosis of acute infection by seroconversion determination through testing acute and convalescent sera
  • Performance characteristics have not been established on children
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Detectable Analytes Mumps IgG
Type Immunosimplicity™ Mumps IgG ELISA Test Kit
Includes Antigen wells, cut-off calibrator, low positive control, negative control, sample B diluent, wash T concentrate 20X, conjugate, substrate HRP, stop N solution
Quantity 96 Tests