Learn More
Diamedix™ Immunosimplicity™ Rubella IgG Test Kit
SDP
Catalog No. 23451023
Encompass
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
23-451-023 96 Tests
1 options
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Catalog No. 23-451-023 Supplier Diamedix™ Supplier No. 720360
Add to Cart
Add to Cart
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes
Antigen Wells, Standards: 0 IU/mL, 10 IU/mL, 50 IU/mL, High Positive Control, Low Positive Control, Negative Control, Sample A Diluent, Wash S Concentrate (20X), Conjugate, Substrate HRP, Stop M Solution
Detects IgG antibodies to rubella in human serum
The Diamedix™ Immunosimplicity™ Rubella IgG Test Kit is a qualitative and quantitative indirect enzyme immunoassay
  • Detects IgG antibodies to rubella in human serum
  • Aids in the assessment of the patient's immunological response to rubella and in the determination of the immune status of individuals, including females of child-bearing age
  • The evaluation of acute and convalescent sera can aid in the diagnosis of current or recent infection with rubella
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Detectable Analytes Rubella IgG
Type Immunosimplicity™ Rubella IgG Test Kit
Includes Antigen wells, standards: 0 IU/mL, 10 IU/mL, 50 IU/mL, high positive control, low positive control, negative control, sample A diluent, wash S concentrate 20X, conjugate, substrate HRP, stop M solution
Quantity 96 Tests