Learn More
MSC Bosch Dust Extractor Hose, 16.4 ft, 35 mm, Plastic, Crush Resistant
Catalog No. 502838445
Encompass
This dust extractor hose is designed for high-performance dust extraction systems. It features a crush-resistant construction and a locking ring for secure connections to power tool adapters and dust collection guards/shrouds. Measuring 16.4 ft in length with a 35 mm diameter, it is suitable for a wide range of dust management applications.
  • Crush-resistant construction for durability
  • Locking ring ensures secure connection to power tool adapters and dust collection guards
  • Flexible design allows for easy maneuverability
  • Compatible with professional dust extractors
  • Includes an adapter for connection to power tools
  • Suitable for use with various tools including planers and sanders
Catalog No. 50-283-8445 Supplier MSC Supplier No. 47824115
This dust extractor hose is designed for high-performance dust extraction systems. It features a crush-resistant construction and a locking ring for secure connections to power tool adapters and dust collection guards/shrouds. Measuring 16.4 ft in length with a 35 mm diameter, it is suitable for a wide range of dust management applications.
  • Crush-resistant construction for durability
  • Locking ring ensures secure connection to power tool adapters and dust collection guards
  • Flexible design allows for easy maneuverability
  • Compatible with professional dust extractors
  • Includes an adapter for connection to power tools
  • Suitable for use with various tools including planers and sanders