Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diamide

Catalog No. 503081500 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1500 500 mg
50-308-1499 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-308-1500 Supplier Sigma Aldrich Supplier No. D36485G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diamide

Specifications

CAS 57757-57-0
Molecular Formula C14H16N2O8S2
Linear Formula C14H16N2O8S2
MDL Number MFCD00042045
Quantity 500 mg
Synonym 1,1'-Azobis(N,N-dimethylformamide); N,N,N',N'-Tetramethylazodicarboxamide; Azodicarboxylic acid bis(dimethylamide); Diazenedicarboxylic acid bis(N,N-dimethylamide); TMAD
Molecular Weight (g/mol) 404.42 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.