Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamide, MP Biomedicals

Catalog No. ICN10152705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10152705 5 g
ICN10152701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10152705 Supplier MP Biomedicals, Inc Supplier No. 0210152705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10465-78-8
Molecular Formula C6H12N4O2
Molecular Weight (g/mol) 172.188
InChI Key VLSDXINSOMDCBK-BQYQJAHWSA-N
Synonym diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
PubChem CID 5353800
ChEBI CHEBI:48963
IUPAC Name (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
SMILES CN(C)C(=O)N=NC(=O)N(C)C

Specifications

Quantity 5 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.