Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamide, MP Biomedicals

Catalog No. MP021015275 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021015275 500 mg
MP021015271 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021015275 Supplier MP Biomedicals, Inc Supplier No. 02101527.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Diamide is used as a thiol oxidizing agent
  • It has also been used to titrate protein glutathiolation to discriminate from other oxidative protein modifications
  • Diamide treatment increased protein glutathiolation in a concentration-dependent manner and had comparably little effect on protein-protein disulfide formation

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10465-78-8
Molecular Formula C6H12N4O2
Molecular Weight (g/mol) 172.188
MDL Number MFCD00008318
InChI Key VLSDXINSOMDCBK-BQYQJAHWSA-N
Synonym diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
PubChem CID 5353800
ChEBI CHEBI:48963
IUPAC Name (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
SMILES CN(C)C(=O)N=NC(=O)N(C)C

Specifications

Melting Point 104°C to 115°C
Color Yellow
Linear Formula C6H12N4O2
Quantity 500 mg
Solubility Information Soluble in water.
Formula Weight 172.19
Physical Form Crystalline
Chemical Name or Material DIAMIDE
Recommended Storage Store at -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.