Learn More
Crescent Chemical Co Inc DIAMIDINO-2-PHENYLINDOLE-2HCL-
SDP
Catalog No. 50247502
Encompass_Preferred
Diamidino-2-phenylindole-2HCl analytical grade Fluorescent dye binding selectively to DNA. For demonstration of mycoplasmas and viruses in cells (1). For fluorescent chromosome staining (2). Dye for brain stem (3). Size - 10MG Storage Conditions - +2 °C TO +8 °C
Catalog No. 50-247-502 Supplier Crescent Chemical Co Inc Supplier No. SE18860.01
Diamidino-2-phenylindole-2HCl analytical grade Fluorescent dye binding selectively to DNA. For demonstration of mycoplasmas and viruses in cells (1). For fluorescent chromosome staining (2). Dye for brain stem (3). Size - 10MG Storage Conditions - +2 °C TO +8 °C