Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diamminedinitritoplatinum(II) Solution

Catalog No. 339881100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-988-1100G 100 g
33-988-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 33-988-1100G Supplier Sigma Aldrich Supplier No. 339881100G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diamminedinitritoplatinum(II) is an active precursor, extensively used in the preparation of Pt-based catalysts on supports such as carbon black and graphene nanosheets. Diamminedinitritoplatinum(II) solution can be used as a platinum source in the synthesis of CoXPt1-X alloy nanowires.

Specifications

CAS 14286-02-3
Density 1.015 g/mL at 25°C
Molecular Formula H6N4O4Pt
Linear Formula Pt(NH3)2(NO2)2
MDL Number MFCD00011622
Quantity 100 g
Molecular Weight (g/mol) 321.15 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.