Learn More
Crescent Chemical Co Inc DIAMINOBENZIDINE-4HCL-XH2O
SDP
Catalog No. 50247505
Encompass_Preferred
Diaminobenzidine-4HCl-xH2O research grade Vials under argon. For histology and ultrahistochemistry, yields osmiophilic oxidation products (1). Demonstration of ultrastructural peroxidase (2). Size - 1G Storage Conditions - +2 °C TO +8 °C Catalog Number - 18865.
Catalog No. 50-247-505 Supplier Crescent Chemical Co Inc Supplier No. SE18865.02
Diaminobenzidine-4HCl-xH2O research grade Vials under argon. For histology and ultrahistochemistry, yields osmiophilic oxidation products (1). Demonstration of ultrastructural peroxidase (2). Size - 1G Storage Conditions - +2 °C TO +8 °C Catalog Number - 18865.