Learn More
Diaminomaleonitrile, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 178.0°C to 179.0°C |
| Color | Brown |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Linear Formula | NCC(NH2)=C(NH2)CN |
| Quantity | 100 g |
| Beilstein | 04, II, 949 |
| Fieser | 05,175; 04,480; 13,327; 01,1256; 02,454; 04,565; 06,657; 07,414; 08,524; 09,512; 10,455; 13,301 |
| Solubility Information | Solubility in water: 5g/L (25°C). Other solubilities: soluble in methanol, ethanol, dmf and dimethyl, sulfoxide, slightly soluble in ethyl ether and dioxane, insol. in benzene,acetone,xylene,toluene and cs2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Harmful if inhaled. Harmful in contact with skin. Contact with acids liberates very toxic gas. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. |
| EINECSNumber | 214-697-6 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.