Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamminepalladium(II) nitrite solution, Pd 8-10% w/w (cont. Pd), Thermo Scientific™

Catalog No. AA11038NJ
Change view
Click to view available options
0.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA11038NJ 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA11038NJ Supplier Thermo Scientific Chemicals Supplier No. 11038NJ

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 28068-05-5
Odor Ammonia-like
Molecular Formula H6N4O4Pd
Quantity 0.5g
MDL Number MFCD00038712
UN Number UN2672
Synonym trans-diamminedinitropalladium ii, diamminepalladium ii nitrite, diamminebis nitrito-o palladium, 2no2.pd.2nh3, dinitritodiamminepalladium ii, azane; palladium 2+ ; dinitrite, diamminepalladium ii nitrite solution, palladium 2+ diamine dinitrite, palladium, diamminebis nitrito-kappao, palladium 2+ ion diamine dinitrite
InChI Key FEVAROJZEMZJGB-UHFFFAOYSA-L
SMILES N.N.N(=O)[O-].N(=O)[O-].[Pd+2]
IUPAC Name azane;palladium(2+);dinitrite
Molecular Weight (g/mol) 232.492
PubChem CID 168754
Formula Weight 232.47 (Anhydrous)
Concentration or Composition (by Analyte or Components) Pd 8 to 10% w/w (cont. Pd)
Chemical Name or Material Diamminepalladium(II) nitrite solution
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMMONIA SOLUTION
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.