Learn More
Medchemexpress LLC Diammonium Glycyrrhizinate | 79165-06-3 | 1 ML
SDP
Catalog No. 5000420200
Encompass_Preferred
Diammonium glycyrrhizinate is a substance extracted and purified from a traditional Chinese medicinal herb. It has anti-inflammatory effects, resistance to biological oxidation, membranous protection, and a weak steroidal action. It protects by downregulating inflammation cytokines, suppressing the NF-κB pathway, and restoring superoxide dismutase. It can be used as a hepatic protector and is studied in research for most liver diseases.
  • Has anti-inflammatory effects
  • Provides resistance to biological oxidation
  • Offers membranous protection
  • Exhibits weak steroidal action
  • Downregulates inflammation cytokines
  • Suppresses the NF-κB pathway
  • Restores superoxide dismutase
  • Can be used as a hepatic protector
  • Studied in research for most liver diseases
Catalog No. 50-004-20200 Supplier Medchemexpress LLC Supplier No. HYN680410MM/1ML
Diammonium glycyrrhizinate is a substance extracted and purified from a traditional Chinese medicinal herb. It has anti-inflammatory effects, resistance to biological oxidation, membranous protection, and a weak steroidal action. It protects by downregulating inflammation cytokines, suppressing the NF-κB pathway, and restoring superoxide dismutase. It can be used as a hepatic protector and is studied in research for most liver diseases.
  • Has anti-inflammatory effects
  • Provides resistance to biological oxidation
  • Offers membranous protection
  • Exhibits weak steroidal action
  • Downregulates inflammation cytokines
  • Suppresses the NF-κB pathway
  • Restores superoxide dismutase
  • Can be used as a hepatic protector
  • Studied in research for most liver diseases