Learn More
Medchemexpress LLC Diammonium glycyrrhizinate | 79165-06-3 | MFCD10566634 | 50 MG
SDP
Catalog No. 5000418029
Encompass_Preferred
Diammonium glycyrrhizinate is a substance extracted and purified from a traditional Chinese medicinal herb. It has anti-inflammatory effects, resistance to biologic oxidation, membranous protection, and a weak steroidal action. It protects by downregulating inflammation cytokines, suppressing the NF-κB pathway, and restoring superoxide dismutase. It can be used as a hepatic protector and is suitable for research on most liver diseases.
  • Anti-inflammatory effects
  • Resistance to biologic oxidation
  • Membranous protection
  • Weak steroidal action
  • Downregulates inflammation cytokines
  • Suppresses the NF-κB pathway
  • Restores superoxide dismutase
  • Suitable for research on most liver diseases
  • Can be used as a hepatic protector
Catalog No. 50-004-18029 Supplier Medchemexpress LLC Supplier No. HYN680450MG
Diammonium glycyrrhizinate is a substance extracted and purified from a traditional Chinese medicinal herb. It has anti-inflammatory effects, resistance to biologic oxidation, membranous protection, and a weak steroidal action. It protects by downregulating inflammation cytokines, suppressing the NF-κB pathway, and restoring superoxide dismutase. It can be used as a hepatic protector and is suitable for research on most liver diseases.
  • Anti-inflammatory effects
  • Resistance to biologic oxidation
  • Membranous protection
  • Weak steroidal action
  • Downregulates inflammation cytokines
  • Suppresses the NF-κB pathway
  • Restores superoxide dismutase
  • Suitable for research on most liver diseases
  • Can be used as a hepatic protector