Learn More
Selleck Chemical LLC Diammonium Glycyrrhizinate S3622-25mg
SDP
Catalog No. 503109267
Encompass_Preferred
Diammonium glycyrrhizinate (DG) a traditional Chinese medicine (TCM) is extracted and purified from liquorices (Radix glycyrrhizae) It is known for its anti-inflammatory effects resistance to biologic oxidation and membranous protection DG is able to reduce inflammatory injury via suppression of NF B TNF and intercellular adhesion molecule 1
Catalog No. 50-310-9267 Supplier Selleck Chemical LLC Supplier No. S362225MG
Diammonium glycyrrhizinate (DG) a traditional Chinese medicine (TCM) is extracted and purified from liquorices (Radix glycyrrhizae) It is known for its anti-inflammatory effects resistance to biologic oxidation and membranous protection DG is able to reduce inflammatory injury via suppression of NF B TNF and intercellular adhesion molecule 1