Learn More
Selleck Chemical LLC Diammonium Glycyrrhizinate S3622-1g
SDP
Catalog No. 503109735
Encompass_Preferred
Diammonium glycyrrhizinate (DG) a traditional Chinese medicine (TCM) is extracted and purified from liquorices (Radix glycyrrhizae) It is known for its anti-inflammatory effects resistance to biologic oxidation and membranous protection DG is able to reduce inflammatory injury via suppression of NF B TNF and intercellular adhesion molecule 1
Catalog No. 50-310-9735 Supplier Selleck Chemical LLC Supplier No. S36221G
Diammonium glycyrrhizinate (DG) a traditional Chinese medicine (TCM) is extracted and purified from liquorices (Radix glycyrrhizae) It is known for its anti-inflammatory effects resistance to biologic oxidation and membranous protection DG is able to reduce inflammatory injury via suppression of NF B TNF and intercellular adhesion molecule 1