Learn More
Medchemexpress LLC Diammonium phosphate | 7783-28-0 | MFCD00010891 | 132.06 g/mol | (NH4)2HPO4 | 1kg
SDP
Catalog No. 5000269988
Encompass_Preferred
Diammonium phosphate is a neutral and efficient catalyst in 1 8 Dioxo octahydroxanthene derivatives synthesis Diammonium phosphate can be used as an excipient such as diuretic buffer effervescent[1][2]
Catalog No. 50-002-69988 Supplier Medchemexpress LLC Supplier No. HYY03021KG
Diammonium phosphate is a neutral and efficient catalyst in 1 8 Dioxo octahydroxanthene derivatives synthesis Diammonium phosphate can be used as an excipient such as diuretic buffer effervescent[1][2]