Learn More
Medchemexpress LLC Diammonium phosphate | 7783-28-0 | MFCD00010891 | 99.1% | 132.06 g/mol | H9N2O4P (alternate: (NH4)2HPO4) | 500g
SDP
Catalog No. 5000268706
Encompass_Preferred
Diammonium phosphate is a neutral and efficient catalyst in 1 8 Dioxo octahydroxanthene derivatives synthesis Diammonium phosphate can be used as an excipient such as diuretic buffer effervescent[1][2]
Catalog No. 50-002-68706 Supplier Medchemexpress LLC Supplier No. HYY0302500G
Diammonium phosphate is a neutral and efficient catalyst in 1 8 Dioxo octahydroxanthene derivatives synthesis Diammonium phosphate can be used as an excipient such as diuretic buffer effervescent[1][2]