Learn More
Electron Microscopy Sciences DiATOME Resharpen Cryo Trim 45° Knife angle Tool
SDP
Catalog No. 501900153
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Catalog No. 50-190-0153 Supplier Electron Microscopy Sciences Supplier No. CTTR
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.