Learn More
Electron Microscopy Sciences Diamond Disc 12" 1500 grit, PSA Steel Magneti Back
SDP
Catalog No. 501929771
Encompass_Preferred
Flexible Diamond Discs will reduce the number of steps in the grinding/polishing process along with a decrease in preparation time. Our Flexible Diamond Discs replace traditional SiC discs for most applications and give a superior finish and perfectly flat specimens. The patterned design minimizes build up of abraded material.
Particles: 1500 grit
Backing: PSA Steel Plate Magnetic
Catalog No. 50-192-9771 Supplier Electron Microscopy Sciences Supplier No. 5029024
Flexible Diamond Discs will reduce the number of steps in the grinding/polishing process along with a decrease in preparation time. Our Flexible Diamond Discs replace traditional SiC discs for most applications and give a superior finish and perfectly flat specimens. The patterned design minimizes build up of abraded material.
Particles: 1500 grit
Backing: PSA Steel Plate Magnetic