Learn More
Electron Microscopy Sciences PremaDisk S-EC Diamond Grinding Disc, 350 mm diameter, 15µm Particles
SDP
Catalog No. 501930287
Encompass_Preferred
These new universal grinding pads for all magnetic systems will replace 200-300 SiC paper discs saving you not only preparation time, but also money. The very high stock removal rate results in fast and clean mounting. Thickness of 0.2 mm with a non-erosion backing. Only water coolant is needed. Replacement for Piano, Cameo, Galaxy, and Apex DGD.
Diameter: 350 mm
Particles: 15µm
Catalog No. 50-193-0287 Supplier Electron Microscopy Sciences Supplier No. 5029212
These new universal grinding pads for all magnetic systems will replace 200-300 SiC paper discs saving you not only preparation time, but also money. The very high stock removal rate results in fast and clean mounting. Thickness of 0.2 mm with a non-erosion backing. Only water coolant is needed. Replacement for Piano, Cameo, Galaxy, and Apex DGD.
Diameter: 350 mm
Particles: 15µm