Learn More
Electron Microscopy Sciences PremaDisk S-EC Diamond Grinding Disc, 250 mm diameter, 6µm Particles
SDP
Catalog No. 501930289
Encompass_Preferred
These new universal grinding pads for all magnetic systems will replace 200-300 SiC paper discs saving you not only preparation time, but also money. The very high stock removal rate results in fast and clean mounting. Thickness of 0.2 mm with a non-erosion backing. Only water coolant is needed. Replacement for Piano, Cameo, Galaxy, and Apex DGD.
Diameter: 250 mm
Particles: 6µm
Catalog No. 50-193-0289 Supplier Electron Microscopy Sciences Supplier No. 5029214
These new universal grinding pads for all magnetic systems will replace 200-300 SiC paper discs saving you not only preparation time, but also money. The very high stock removal rate results in fast and clean mounting. Thickness of 0.2 mm with a non-erosion backing. Only water coolant is needed. Replacement for Piano, Cameo, Galaxy, and Apex DGD.
Diameter: 250 mm
Particles: 6µm