Learn More
Electron Microscopy Sciences PremaDisk Diamond Grinding Disc SM Medium-Hard Bond, 6 µm Particles, 250 mm diameter
SDP
Catalog No. 501929930
Encompass_Preferred
PremaDisk Flexible Diamond Grinding Discs offer superior surface finishes that are usually only seen in lapping processes.
The SM disc is a medium-hard resin bond base for fine grinding and polishing of metals, ceramics and glass.
The SH disc is a hard resin bond base for applications where a very sharp edge must be maintained on the workpiece.
Both discs increase productivity by combining optimum surface quality, removal rates, and tool life in the same process.
Particles: 6 µm
Diameter: 250 mm
Catalog No. 50-192-9930 Supplier Electron Microscopy Sciences Supplier No. 5029314
PremaDisk Flexible Diamond Grinding Discs offer superior surface finishes that are usually only seen in lapping processes.
The SM disc is a medium-hard resin bond base for fine grinding and polishing of metals, ceramics and glass.
The SH disc is a hard resin bond base for applications where a very sharp edge must be maintained on the workpiece.
Both discs increase productivity by combining optimum surface quality, removal rates, and tool life in the same process.
Particles: 6 µm
Diameter: 250 mm