Learn More
Electron Microscopy Sciences DiATOME Cryo Diamond Knife Resharpen Dry 3.0 mm 25° Knife angle
SDP
Catalog No. 501898805
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 3.0 mm
Knife angle: 25°
Catalog No. 50-189-8805 Supplier Electron Microscopy Sciences Supplier No. 30CD25R
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 3.0 mm
Knife angle: 25°