Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife Diamond Knife Dry, 2.7 mm, 35° Knife angle
SDP
Catalog No. 501902420
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 2.7 mm
Knife angle: 35°
Catalog No. 50-190-2420 Supplier Electron Microscopy Sciences Supplier No. 27ULD
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 2.7 mm
Knife angle: 35°