Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife 1.5 mm, 45° Knife angle
SDP
Catalog No. 501898722
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 1.5 mm
Knife angle: 45°
Catalog No. 50-189-8722 Supplier Electron Microscopy Sciences Supplier No. 15US
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 1.5 mm
Knife angle: 45°