Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife 3.0 mm, 45° Knife angle
SDP
Catalog No. 501898820
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 3.0 mm
Knife angle: 45°
Catalog No. 50-189-8820 Supplier Electron Microscopy Sciences Supplier No. 30US
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 3.0 mm
Knife angle: 45°