Learn More
Electron Microscopy Sciences Write on Marker Pens, 1/EA
SDP
Catalog No. 502091003
Encompass_Preferred
These pens are made from specially formulated ink and are to be used on anterior surfaces as well as sides of histology cassettes. The ink will dry almost instantly and withstands temperatures up to 195 degree C and is resistant to solvents.
Catalog No. 50-209-1003 Supplier Electron Microscopy Sciences Supplier No. 7210601A
These pens are made from specially formulated ink and are to be used on anterior surfaces as well as sides of histology cassettes. The ink will dry almost instantly and withstands temperatures up to 195 degree C and is resistant to solvents.