Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diamond

Catalog No. 6364445G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-644-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 63-644-45G Supplier Sigma Aldrich Supplier No. 6364445G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diamond nanopowder (DNPs) may be produced by multicathode direct current plasma chemical vapor deposition and high pressure high temperature (HPHT).Potential uses of nano-diamond are in biosensor applications. Surface modification of nano-diamond may enhance its linking mechanism with specific biomolecules. DNPs can be subjected to functionalization post Fenton treatment. These functionalized DNPs may be used as gene delivery systems.

Specifications

CAS 7782-40-3
Density 3.5 g/mL (at 25°C)
Molecular Formula C
Linear Formula C
Quantity 5 g
MDL Number MFCD00211867
Molecular Weight (g/mol) 12.01
Percent Purity ≥95% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.