Learn More
Electron Microscopy Sciences Diamond Disc 10" 400 grit, PSA backing
SDP
Catalog No. 501929716
Encompass_Preferred
Flexible Diamond Discs will reduce the number of steps in the grinding/polishing process along with a decrease in preparation time. Our Flexible Diamond Discs replace traditional SiC discs for most applications and give a superior finish and perfectly flat specimens. The patterned design minimizes build up of abraded material.
Particles: 400 grit
Backing: PSA
Catalog No. 50-192-9716 Supplier Electron Microscopy Sciences Supplier No. 5027017
Flexible Diamond Discs will reduce the number of steps in the grinding/polishing process along with a decrease in preparation time. Our Flexible Diamond Discs replace traditional SiC discs for most applications and give a superior finish and perfectly flat specimens. The patterned design minimizes build up of abraded material.
Particles: 400 grit
Backing: PSA