Learn More
Electron Microscopy Sciences Diamond Disc 12" 120 grit, PSA backing
SDP
Supplier:  Electron Microscopy Sciences 5027009
Encompass_Preferred
Flexible Diamond Discs will reduce the number of steps in the grinding/polishing process along with a decrease in preparation time. Our Flexible Diamond Discs replace traditional SiC discs for most applications and give a superior finish and perfectly flat specimens. The patterned design minimizes build up of abraded material.
Particles: 120 grit
Backing: PSA
Catalog No. 50-192-9708