Learn More
Electron Microscopy Sciences Diamond Disc 12" 320 grit, PSA Steel Plate Back
SDP
Catalog No. 501929738
Encompass_Preferred
Flexible Diamond Discs will reduce the number of steps in the grinding/polishing process along with a decrease in preparation time. Our Flexible Diamond Discs replace traditional SiC discs for most applications and give a superior finish and perfectly flat specimens. The patterned design minimizes build up of abraded material.
Particles: 320 grit
Backing: PSA Steel Plate
Catalog No. 50-192-9738 Supplier Electron Microscopy Sciences Supplier No. 5028015
Flexible Diamond Discs will reduce the number of steps in the grinding/polishing process along with a decrease in preparation time. Our Flexible Diamond Discs replace traditional SiC discs for most applications and give a superior finish and perfectly flat specimens. The patterned design minimizes build up of abraded material.
Particles: 320 grit
Backing: PSA Steel Plate