Learn More
Electron Microscopy Sciences Nano Diamond Dissecting Knife Plastic Handle, Round Knife
SDP
Catalog No. 501931192
Encompass_Preferred
These blades are the finest and sharpest blades for all of your micro-surgical needs. The knives are made from a polychrystalline CVD blade and are plasma sharpened to 0.15 mm thick.
Type: Round Knife
Width: 2.0 mm
Length: 4.0 mm
Thickness: 0.15 mm
Catalog No. 50-193-1192 Supplier Electron Microscopy Sciences Supplier No. 7200520
These blades are the finest and sharpest blades for all of your micro-surgical needs. The knives are made from a polychrystalline CVD blade and are plasma sharpened to 0.15 mm thick.
Type: Round Knife
Width: 2.0 mm
Length: 4.0 mm
Thickness: 0.15 mm