Learn More
Electron Microscopy Sciences Meyco Diamond Dissecting Knife, Type L, Lancet Blade, 0.50 mm thick
SDP
Supplier:  Electron Microscopy Sciences 72026
Encompass_Preferred
Our ultra-sharp diamond blade cuts with minimum pressure, without tearing or damaging the specimen and reduces the risk of specImen distortion. The UNIQUE PRESSURE MECHANISM allows blade actuation with one hand. The knives are of the same quality that DIATOME customers have become accustomed. Distinctive designs combined with DIATOME'S expertise in diamond technology and manufacturing assures you the best quality knife and being able to use it for many years.
Blade: Lancet
Thickness: 0.50 mm
Catalog No. 50-949-880