Learn More
Electron Microscopy Sciences Meyco Diamond Dissecting Knife, Type M-M, Mini-angled Blade, 0.20 mm thick
SDP
Catalog No. 50949851
Encompass_Preferred
Our ultra-sharp diamond blade cuts with minimum pressure, without tearing or damaging the specimen and reduces the risk of specImen distortion. The UNIQUE PRESSURE MECHANISM allows blade actuation with one hand. The knives are of the same quality that DIATOME customers have become accustomed. Distinctive designs combined with DIATOME'S expertise in diamond technology and manufacturing assures you the best quality knife and being able to use it for many years. For extremely fine work .
Blade: Mini-angled
Thickness: 0.20 mm
Catalog No. 50-949-851 Supplier Electron Microscopy Sciences Supplier No. 72025
Our ultra-sharp diamond blade cuts with minimum pressure, without tearing or damaging the specimen and reduces the risk of specImen distortion. The UNIQUE PRESSURE MECHANISM allows blade actuation with one hand. The knives are of the same quality that DIATOME customers have become accustomed. Distinctive designs combined with DIATOME'S expertise in diamond technology and manufacturing assures you the best quality knife and being able to use it for many years. For extremely fine work .
Blade: Mini-angled
Thickness: 0.20 mm