Learn More
Electron Microscopy Sciences Diamond Finishing Disc, Extra Coarse Grit
SDP
Catalog No. 50996839
Encompass_Preferred
Diamond Finishing discs offer extremely aggressive grinding action, yet leave an exceptionally smooth finish. Discs have pressure sensitive adhesive backing for a secure fit on holder and 3/4" (19.1 mm) diameter.
Grit: Extra Coarse
Color: Purple
Catalog No. 50-996-839 Supplier Electron Microscopy Sciences Supplier No. 6214610
Diamond Finishing discs offer extremely aggressive grinding action, yet leave an exceptionally smooth finish. Discs have pressure sensitive adhesive backing for a secure fit on holder and 3/4" (19.1 mm) diameter.
Grit: Extra Coarse
Color: Purple