Learn More
Supelco Inc Diamond Glass Cutter, Wooden Handle
Catalog No. NC9332189
Encompass
Diamond Glass Cutter, Scores/breaks single or double-strength glass, Nickel-plated brass cutter with wooden handle
Catalog No. NC9332189 Supplier Supelco Inc Supplier No. Z1690641EA
Diamond Glass Cutter, Scores/breaks single or double-strength glass, Nickel-plated brass cutter with wooden handle