Learn More
Electron Microscopy Sciences Diamond Glass Cutter
SDP
Catalog No. 5024679
Encompass_Preferred
Metal parts are made from nickel-plated brass, set into a maple handle. The rack can be used to crack single or double strength glass.
Catalog No. 50-246-79 Supplier Electron Microscopy Sciences Supplier No. 70038
Metal parts are made from nickel-plated brass, set into a maple handle. The rack can be used to crack single or double strength glass.