Learn More
Polysciences, Inc. Glass Scriber Diamond 1 scriber Useful for scoring glass strips prior to hand making glass knives marking slides and property identification
SDP
Catalog No. NC9635378
Encompass
Glass Scriber Diamond 1 scriber Useful for scoring glass strips prior to hand making glass knives marking slides and property identification
Catalog No. NC9635378 Supplier Polysciences, Inc. Supplier No. 036361
Glass Scriber Diamond 1 scriber Useful for scoring glass strips prior to hand making glass knives marking slides and property identification