Learn More
Electron Microscopy Sciences Diamond Grinding Disc, 3 µm, 8" diameter
SDP
Catalog No. 50997240
Encompass_Preferred
Dialok Diamond Discs consists of a flat, stainless steel backing with sharp, blocky diamond particles electroformed onto the disc. The special hard nickel alloy that is used in Dialok is bonded rigidly and it locks the diamond particles in the disc thus providing fast cutting of all hard materials. The sharp particles cut cleanly with no edge rounding. The discs are supplied with pressure sensitive adhesive backing (PSA).
Particles: 3 µm
Diameter: 8"
Catalog No. 50-997-240 Supplier Electron Microscopy Sciences Supplier No. 5034530
Dialok Diamond Discs consists of a flat, stainless steel backing with sharp, blocky diamond particles electroformed onto the disc. The special hard nickel alloy that is used in Dialok is bonded rigidly and it locks the diamond particles in the disc thus providing fast cutting of all hard materials. The sharp particles cut cleanly with no edge rounding. The discs are supplied with pressure sensitive adhesive backing (PSA).
Particles: 3 µm
Diameter: 8"